Skip to Content
Merck
All Photos(1)

Key Documents

W800036

Sigma-Aldrich

5-Hexen-2-one

≥98%

Synonym(s):

Allylacetone

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3COCH2CH2CH=CH2
CAS Number:
Molecular Weight:
98.14
Beilstein:
1699137
MDL number:
UNSPSC Code:
12164502
Flavis number:
07.162
NACRES:
NA.21
food allergen:
no known allergens

reg. compliance

FDA 21 CFR 117

Assay

≥98%

refractive index

n20/D 1.419 (lit.)

bp

128-129 °C (lit.)

density

0.847 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

no known allergens

Organoleptic

ethereal; floral

SMILES string

CC(=O)CCC=C

InChI

1S/C6H10O/c1-3-4-5-6(2)7/h3H,1,4-5H2,2H3

InChI key

RNDVGJZUHCKENF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Disclaimer

For R&D or non-EU Food use. Not for retail sale.

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

75.2 °F - closed cup

Flash Point(C)

24 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service