Skip to Content
Merck
All Photos(1)

Documents

W324300

Sigma-Aldrich

(E)-β-Damascone

≥95%, sum of isomers, FG

Synonym(s):

(E)-1-(2,6,6-Trimethyl-1-cyclohexenyl)-2-buten-1-one

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H20O
CAS Number:
Molecular Weight:
192.30
FEMA Number:
3243
Beilstein:
2046078
EC Number:
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
07.224
NACRES:
NA.21

grade

FG
Fragrance grade
Halal
Kosher

Agency

follows IFRA guidelines

reg. compliance

EU Regulation 1223/2009
EU Regulation 1334/2008 & 178/2002
FDA 21 CFR 117

Assay

≥95%

composition

contains IFRA restricted β-Damascone

density

0.934 g/mL at 20 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

Damascone, cis-β-damascone

Organoleptic

leather; woody; minty; floral; fruity; sweet

SMILES string

C\C=C\C(=O)C1=C(C)CCCC1(C)C

InChI

1S/C13H20O/c1-5-7-11(14)12-10(2)8-6-9-13(12,3)4/h5,7H,6,8-9H2,1-4H3/b7-5+

InChI key

BGTBFNDXYDYBEY-FNORWQNLSA-N

Looking for similar products? Visit Product Comparison Guide

General description

β-Damascone belongs to a family of fragrant products called rose ketones. Despite having a low concentration, beta-Damascone contributes to the rose aroma. It is an important fragrance chemical that is used in perfumery .

Application

β-Damascone is:
  • Used as a precursor to synthesize β-Damascenone, its main contribution to the floral aromas of many wine varieties, as well as in the perfume and flavoring industries .
  • One of the key volatile compounds contributing to the floral aroma of green tea .

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Skin Irrit. 2 - Skin Sens. 1B

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

212.0 °F - closed cup

Flash Point(C)

100 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

New Straightforward Synthesis of ??Damascenone and ??Damascone Derivatives from ??Ionone via Retro???ionol
Campagnole M, and Delmond B
Synthetic Communications, 37(7), 1077-1090 (2007)
Revelation of volatile contributions in green teas with different aroma types by GC-MS and GC-IMS
Liu N, et al.
Food Research International, 112845-112845 (2023)
Synthesis of d6-deuterated analogues of aroma molecules-?-damascenone, ?-damascone and safranal
Mosaferi S, et al.
Results in Chemistry, 4, 100264-100264 (2022)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service