Key Documents
SYNM431005
Synthware all-glass vacuum/inert gas manifold with hollow high vacuum stopcocks
number of ports, 5, Hose Connections: Front-left, Rear-left-right
Synonym(s):
Synthware M431005, vacuum manifold, vacuum unit
About This Item
UNSPSC Code:
40161501
NACRES:
NB.51
Pricing and availability is not currently available.
feature
number of ports 5
packaging
pkg of 1 ea
manufacturer/tradename
Synthware M431005
General description
With 4 mm double oblique, high vacuum hollow glass stopcocks precision ground with corresponding numbers on barrel and plug. Lower vacuum chambers on stopcock are evacuated with a separate hole in the plug in order to hold the plug in the stopcock barrels. Beams are 22 mm O.D. with front-left and rear-left-right 10 mm hose connections. Distance between stopcocks is 75 mm center to center.
Legal Information
Synthware is a trademark of Kemtech America Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)