SYNF601002C
Synthware™ Buchner funnel
2 mL, porosity: coarse
Synonym(s):
Synthware F601002C, filtering funnel
All Photos(1)
About This Item
UNSPSC Code:
41121809
NACRES:
NB.51
Recommended Products
packaging
pkg of 1 ea
manufacturer/tradename
Synthware F601002C
capacity
2 mL
2 mL
Related Categories
General description
With sealed-in glass disc of the listed porosity. Not recommended for filtration where pressure would exceed 15psi. XC-extra coarse, C-Coarse, M-Medium, F-Fine porosity.
Legal Information
Synthware is a trademark of Kemtech America Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)