Key Documents
SYNF561025
Synthware reaction tube with high vacuum valve
25 mL, joint: ST/NS 14/20, valve size 0-4 mm
Synonym(s):
Synthware F561025, Schlenk flask
About This Item
UNSPSC Code:
41121700
NACRES:
NB.51
Pricing and availability is not currently available.
material
colorless tube
glass tube
Joint
joint: ST/NS14/20
sterility
non-sterile
feature
graduations: no
Push fit
packaging
pkg of 1 ea
manufacturer/tradename
Kemtech F561025
capacity
25 mL
25 mL
valve size
0-4 mm
General description
Supplied with high vacuum valve with PTFE o-ring at tip and PTFE wiper (“umbrella”) to protect Viton® O-ring on shaft.
Legal Information
Synthware is a trademark of Kemtech America Inc.
Viton is a registered trademark of E. I. du Pont de Nemours and Company
Choose from one of the most recent versions:
Certificates of Analysis (COA)