Skip to Content
Merck
All Photos(1)

Key Documents

SYNB257040L

Synthware mineral oil bubbler

O.D. × H 500 mm

Synonym(s):

Synthware B257040L

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121800
NACRES:
NB.51

packaging

pkg of 1 ea

manufacturer/tradename

Synthware B257040L

O.D. × H

500 mm

Looking for similar products? Visit Product Comparison Guide

General description

With 10 mm O.D. hose connection.

Legal Information

Synthware is a trademark of Kemtech America Inc.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service