Skip to Content
Merck
All Photos(1)

Key Documents

QBD11221

Sigma-Aldrich

Bromoacetamido-dPEG ®4-amido-DBCO

Synonym(s):

Polyethylene glycol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C31H38BrN3O7
Molecular Weight:
644.55
UNSPSC Code:
12352101
NACRES:
NA.22
Pricing and availability is not currently available.

Assay

>90%

form

solid or viscous liquid

reaction suitability

reagent type: cross-linking reagent
reaction type: click chemistry

functional group

bromo

polymer architecture

shape: linear
functionality: heterobifunctional

shipped in

ambient

storage temp.

−20°C

SMILES string

BrCC(NCCOCCOCCOCCOCCC(NCCC(N1C(C=CC=C2)=C2C#CC(C=CC=C3)=C3C1)=O)=O)=O

InChI key

WWMVSWYAKQXVAU-UHFFFAOYSA-N

Features and Benefits

Bromoacetamido-dPEG®₄-amido-DBCO is a crosslinker with a bromoacetamido which can react with a free thiol and a DBCO which can be used for a copper-free "click" chemistry reaction with an azide group.

Legal Information

Products Protected under U.S. Patents # 7,888,536 B2
dPEG is a registered trademark of Quanta BioDesign

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service