All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C46H68N2O18
CAS Number:
Molecular Weight:
937.03
MDL number:
UNSPSC Code:
12352108
NACRES:
NA.22
Recommended Products
Assay
>90%
form
solid or viscous liquid
reaction suitability
reaction type: Pegylations
reagent type: cross-linking reagent
functional group
Fmoc
polymer architecture
shape: linear
functionality: heterobifunctional
shipped in
ambient
storage temp.
−20°C
SMILES string
O=C(OCC1C2=C(C3=C1C=CC=C3)C=CC=C2)NCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCC(ON4C(CCC4=O)=O)=O
Features and Benefits
Fmoc-N-amido-dPEG ®₁₂-NHS ester can be used to introduce a dPEG linker and NHS ester during peptide synthesis.
Legal Information
Products Protected under U.S. Patents # 7,888,536 B2
dPEG is a registered trademark of Quanta BioDesign
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service