Key Documents
HAM201150
Hamilton® Gastight® syringe, general use, 1700 series
model, 1750, AccuDil (AD) end, volume 500 μL
About This Item
UNSPSC Code:
41122001
NACRES:
NB.03
Pricing and availability is not currently available.
material
Borosilicate glass/PTFE
Gastight syringe
feature
autoclavable: no
model 1750, AccuDil (AD) end
manufacturer/tradename
Hamilton
parameter
20-50 °C temp. range (68-122 °F)
700 psig pressure
volume
500 μL
Related Categories
Legal Information
GASTIGHT is a registered trademark of Hamilton Co.
Hamilton is a registered trademark of Hamilton Co.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Questions
  1. Can I use this syringe to collect phosphine (for bioefficacy studies on insects)
    1 answer
    1. The 1700 series is a gas-tight syringe used for manual and automated gas chromatography and high-performance liquid chromatography injections. It is also used for applications requiring accurate measuring and dispensing of liquid.
      Helpful?