Skip to Content
Merck
All Photos(1)

Key Documents

H1006

Sigma-Aldrich

Heptafluorobutyric anhydride

≥98%

Synonym(s):

HFAA, HFBA, Perfluorobutyric anhydride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CF3CF2CF2CO)2O
CAS Number:
Molecular Weight:
410.06
Beilstein:
856036
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥98%

form

liquid

refractive index

n20/D 1.287 (lit.)

bp

108-110 °C (lit.)

mp

−43 °C (lit.)

density

1.674 g/mL at 20 °C (lit.)

storage temp.

room temp

SMILES string

FC(F)(F)C(F)(F)C(F)(F)C(=O)OC(=O)C(F)(F)C(F)(F)C(F)(F)F

InChI

1S/C8F14O3/c9-3(10,5(13,14)7(17,18)19)1(23)25-2(24)4(11,12)6(15,16)8(20,21)22

InChI key

UFFSXJKVKBQEHC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Heptafluorobutyric anhydride can be used as a derivatizing agent for the determination of:
  • Alkylphenol ethoxylates and brominated flame retardants in sewage sludge samples.
  • Free amino acids in sugarcane by GC analysis.

Packaging

5ML and 25ML in glass bottle

10 AMP in ampule

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

An integrated method for the simultaneous determination of alkylphenol ethoxylates and brominated flame retardants in sewage sludge samples by ultrasonic-assisted extraction, solid phase clean-up, and GC-MS analysis
Chokwe T, et al.
Microchemical Journal, Devoted to the Application of Microtechniques in All Branches of Science, 123, 230-236 (2015)
Anne Bohin et al.
Analytical biochemistry, 340(2), 231-244 (2005-04-21)
In previous articles [Anal. Biochem. 284 (2000) 201; J. Lipid Res. 43 (2002) 794], we reported that the GC/MS identification and quantification of nearly all constituents of glycolipids could be obtained on the same sample in a single GC/MS analysis
O Beck et al.
Journal of analytical toxicology, 22(1), 45-49 (1998-03-10)
The use of mushrooms containing the hallucinogenic substance psilocybin for intentional intoxication is relatively common. Occasionally, this results in adverse reactions with typical tachycardia that is not evidently caused by psilocybin. This study demonstrates the presence of phenylethylamine in the
Ronald Agius et al.
Forensic science international, 196(1-3), 3-9 (2010-01-12)
The analysis of ethyl glucuronide (EtG) in hair is a powerful tool for chronic alcohol abuse control because of the typical wide detection window of the hair matrix and due to the possibility of segmentation, allowing evaluation of alcohol consumption
C Fakt et al.
Journal of chromatography. B, Biomedical sciences and applications, 700(1-2), 93-100 (1997-12-09)
A convenient and sensitive method for the quantitative determination of poly(ethylene glycol) 400 in plasma and urine with capillary gas chromatography-mass spectrometry has been developed. The sample preparation involves solid-phase extraction with subsequent derivatization with heptafluorobutyric anhydride, which proved to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service