Skip to Content
Merck
All Photos(1)

Key Documents

GF50025338

Polytetrafluoroethylene

(PTFE), powder, mean particle size 6-9micron, weight 100 g

Synonym(s):

FP306010

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352100
NACRES:
NA.23
Pricing and availability is not currently available.

form

powder

manufacturer/tradename

Goodfellow 500-253-38

weight

100 g

General description

For updated SDS information please visit www.goodfellow.com

Application

  • Nanocomposites based on polytetrafluoroethylene and ultrahigh molecular weight polyethylene: This review discusses the enhancements in mechanical strength and wear resistance of equipment used in polar conditions through the use of PTFE nanocomposites (IV Kirillina, LA Nikiforov, AA Okhlopkova, 2014).
  • Structure and properties of polytetrafluoroethylene (PTFE) fibers: This study characterizes PTFE fibers using SEM and FT-IR, highlighting their structural properties and potential for diverse applications (R Wang, G Xu, Y He, 2017).
  • Manufacturing technology of composite materials: Explores the modification techniques of PTFE-based polymer composites, aiming to improve their wear resistance and technological applications (A Panda, K Dyadyura, J Valíček, M Harničárová, 2017).
  • XPS and FTIR studies of polytetrafluoroethylene thin films: Analyzes the chemical structure of PTFE thin films, enhancing understanding of their properties and applications in various industries (J Piwowarczyk, R Jędrzejewski, D Moszyński, 2019).
  • High-density polytetrafluoroethylene membranes in guided bone and tissue regeneration procedures: Reviews the use of PTFE membranes in medical applications, particularly for guided bone and tissue regeneration, comparing expanded vs. dense PTFE forms (JM Carbonell, IS Martín, A Santos, A Pujol, 2014).

Legal Information

Product of Goodfellow

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service