GF40537065
Nickel silver (18) alloy, Cu65Ni18Zn17
1000mm tube, 1.0mm outside diameter, 0.1mm wall thickness, 0.8mm inside diameter, hard
Synonym(s):
CZ057120
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
11101711
NACRES:
NA.23
form
tubes
manufacturer/tradename
Goodfellow 405-370-65
L
1000 mm
O.D. × I.D. × thickness
1.0 mm × 0.8 mm × 0.1 mm
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
For updated SDS information please visit www.goodfellow.com.
Product of Goodfellow
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service