DWKW242839
Wheaton® Dropping Bottle
white bottle, capacity 120 mL
Synonym(s):
W242839
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41121800
NACRES:
NB.17
Recommended Products
material
white bottle
packaging
pkg of 100 ea
manufacturer/tradename
WHEATON® W242839
capacity
120 mL
Looking for similar products? Visit Product Comparison Guide
Related Categories
Application
Opthalmic, IVD Kits, Other
Features and Benefits
- Bottle made of natural or white LDPE (Low Density Polyethylene
- White LDPE bottle helps protect UV light sensitive samples
- Dropper tip caps and tips sold separately
- Packaged in cases of 100 or 1000
Legal Information
WHEATON is a registered trademark of Wheaton Industries, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service