DWK73785-10
KIMBLE® centrifuge tube
with screw-top finish, conical bottom clear glass, capacity (10 mL), without cap, non-sterile
All Photos(1)
About This Item
UNSPSC Code:
41121703
NACRES:
NB.11
Recommended Products
material
conical bottom clear glass
conical bottom
transparent borosilicate glass tube
sterility
non-sterile
feature
without cap
packaging
case of 125 ea Modular trays are shrink-wrapped
manufacturer/tradename
Kimble® 73785-10
capacity
(10 mL)
fitting
15-415 neck for thread
Related Categories
General description
Screw Thread Disposable Centrifuge Tubes
- These tubes are suitable for tissue culture work and general bacteriological use.
- Supplied without caps
- No marking spot
- Manufactured from 51 expansion borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class B requirements
Legal Information
KIMBLE is a registered trademark of DWK Life Sciences
related product
Product No.
Description
Pricing
Choose from one of the most recent versions:
Certificates of Analysis (COA)