DWK73665-18
KIMBLE® KIM-KAP® push cap
blue polypropylene cap, cap diam. (18 mm)
All Photos(1)
About This Item
UNSPSC Code:
41121800
NACRES:
NB.17
Recommended Products
material
blue polypropylene cap
sterility
non-sterile
packaging
case of 1000 ea
manufacturer/tradename
Kimble® 73665-18
cap diam.
(18 mm)
Related Categories
General description
- KIM-KAP polypropylene closures for culture tubes
- Autoclavable
- Color coded
Features and Benefits
- Autoclavable
- Color coded
Legal Information
KIM-KAP is a registered trademark of DWK LIFE SCIENCES LLC
KIMBLE is a registered trademark of DWK Life Sciences
Choose from one of the most recent versions:
Certificates of Analysis (COA)