Skip to Content
Merck
All Photos(3)

Documents

D205001

Sigma-Aldrich

9,10-Diphenylanthracene

97%

Synonym(s):

Anthracene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H18
CAS Number:
Molecular Weight:
330.42
Beilstein:
1914010
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Assay

97%

form

powder

mp

245-248 °C (lit.)

SMILES string

c1ccc(cc1)-c2c3ccccc3c(-c4ccccc4)c5ccccc25

InChI

1S/C26H18/c1-3-11-19(12-4-1)25-21-15-7-9-17-23(21)26(20-13-5-2-6-14-20)24-18-10-8-16-22(24)25/h1-18H

InChI key

FCNCGHJSNVOIKE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application


  • Achieving spin-triplet exciton transfer between silicon and molecular acceptors for photon upconversion.: This research demonstrates the transfer of triplet excitons from silicon to 9,10-diphenylanthracene, a key mechanism for photon upconversion technologies that could revolutionize solar energy harvesting and optoelectronic devices (Xia et al., 2020).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jin-Ming Lin et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 72(1), 126-132 (2008-11-11)
The decomposition of peroxymonocarbonate (HCO(4)(-)) has been investigated by flow-injection chemiluminescence (CL) method. An ultraweak CL was observed during mixing the bicarbonate and hydrogen peroxide solution in organic cosolvent. An appropriate amount of fluorescent organic compounds, such as dichlorofluorescein (DCF)
Weibin Cui et al.
Chemical communications (Cambridge, England), (8)(8), 1017-1019 (2008-02-20)
The first soluble conjugated poly(2,6-anthrylene) with 9,10-diphenyl-anthracene as the repeating unit is reported; photophysical studies reveal that this polymer represents a novel well-conjugated system.
Jin-Ming Lin et al.
The journal of physical chemistry. B, 112(26), 7850-7855 (2008-06-12)
In this work, an online preparation of peroxymonocarbonate was formed innovatively, which offered the reliable intermediate for further investigation. Gold colloids with nanoparticles of different sizes were found to enhance the chemiluminescence (CL) of the peroxymonocarbonate-eosin Y system, and the
[Synthesis of novel fluorescent 9,10-bisphenylethynylanthracene-derived pseudonucleoside and its introduction into oligonucleotides].
A D Malakhov et al.
Bioorganicheskaia khimiia, 25(12), 933-937 (2000-03-29)
Regression analysis of electrochemical data with expanding space grid digital simulation at spherical electrodes.
J V Arena et al.
Analytical chemistry, 58(7), 1481-1488 (1986-06-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service