Skip to Content
Merck
All Photos(1)

Key Documents

D109207

Sigma-Aldrich

2,5-Dihydroxy-1,4-benzenediacetic acid

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(HO)2C6H2(CH2CO2H)2
CAS Number:
Molecular Weight:
226.18
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Assay

97%

form

powder or crystals

mp

281-283 °C (dec.) (lit.)

SMILES string

OC(=O)Cc1cc(O)c(CC(O)=O)cc1O

InChI

1S/C10H10O6/c11-7-1-5(3-9(13)14)8(12)2-6(7)4-10(15)16/h1-2,11-12H,3-4H2,(H,13,14)(H,15,16)

InChI key

MCLKERLHVBEZIW-UHFFFAOYSA-N

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
663158264474D109401
Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

100

mp

281-283 °C (dec.) (lit.)

mp

202 °C (dec.)

mp

115-118 °C (lit.)

mp

208-211 °C (dec.) (lit.)

form

powder or crystals

form

solid

form

solid

form

powder

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service