BR155540
BRAND® weighing dish
polystyrene, L × W × H 45 mm × 45 mm × 7 mm, square
Synonym(s):
Dishes, Weighing dishes
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41121812
NACRES:
NB.71
Recommended Products
material
polystyrene
square
packaging
case of 500 ea
manufacturer/tradename
BRAND 155540
L × W × H
45 mm × 45 mm × 7 mm
dish L × W
45 mm × 45 mm
volume
7 mL
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
The BRAND® weighing dish is made from impact-resistant polystyrene (PS) and features an antistatic design, making it ideal for accurate weighing in laboratory settings. Its light and flat design ensures easy handling and stability during use. With a wall thickness of approximately 0.2 mm, this weighing dish provides a reliable surface for measuring various substances. The antistatic properties help prevent the adherence of dust and particles, ensuring clean and precise measurements. This weighing dish is an essential tool for any laboratory focused on efficiency and accuracy in weighing applications.
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service