Skip to Content
Merck
All Photos(2)

Key Documents

ALD00596

Sigma-Aldrich

N-((1S,2S)-1-(3,5-Di-tert-butylphenyl)-2-(quinolin-2-yl)butyl)acetamide

≥95%

Synonym(s):

Chiral acetyl-protected aminoethyl quinoline ligand, Et-APAQ

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C29H38N2O
Molecular Weight:
430.62
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.22

Quality Level

Assay

≥95%

form

powder or crystals

reaction suitability

reaction type: C-C Bond Formation
reagent type: catalyst
reagent type: ligand
reaction type: C-H Activation

SMILES string

N([C@@H]([C@H](CC)c2nc3c(cc2)cccc3)c1cc(cc(c1)C(C)(C)C)C(C)(C)C)C(=O)C

InChI

1S/C29H38N2O/c1-9-24(26-15-14-20-12-10-11-13-25(20)31-26)27(30-19(2)32)21-16-22(28(3,4)5)18-23(17-21)29(6,7)8/h10-18,24,27H,9H2,1-8H3,(H,30,32)/t24-,27-/m1/s1

InChI key

NZPXTQVMJQHEBG-SHQCIBLASA-N

Application

This chiral acetyl-protected aminoethyl quinoline (APAQ) ligand was developed by the Yu group for ligand-enabled acceleration of Pd-catalyzed enantioselective β-methylene C–H functionalization, which is important for the generation of β-chiral centers in asymmetric synthesis.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

Yu program focuses on efficient C–H bond activation for drug synthesis, using simple starting materials.

Yu program focuses on efficient C–H bond activation for drug synthesis, using simple starting materials.

Yu program focuses on efficient C–H bond activation for drug synthesis, using simple starting materials.

Yu program focuses on efficient C–H bond activation for drug synthesis, using simple starting materials.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service