Skip to Content
Merck
All Photos(3)

Key Documents

A75706

Sigma-Aldrich

2-Amino-6-methylpyridine

98%

Synonym(s):

6-Amino-2-picoline

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H8N2
CAS Number:
Molecular Weight:
108.14
Beilstein:
107048
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

crystals

bp

208-209 °C (lit.)

mp

40-44 °C (lit.)

SMILES string

Cc1cccc(N)n1

InChI

1S/C6H8N2/c1-5-3-2-4-6(7)8-5/h2-4H,1H3,(H2,7,8)

InChI key

QUXLCYFNVNNRBE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Dermal - Acute Tox. 3 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

217.4 °F - closed cup

Flash Point(C)

103 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Vidya V M et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 245, 118940-118940 (2020-09-27)
The synthesis of five D-n-A type star-shaped octupolar molecules is presented in the current work. The exploration of the potential applicability of molecules under study in organic optoelectronics as electron or hole transporting materials is carried out by DFT methods.
Mohanad Darawsheh et al.
European journal of medicinal chemistry, 82, 152-163 (2014-06-07)
Starting from the precursor [Zinc Valproate complex] (1), new mixed ligand zinc(II) complexes of valproic acid and nitrogen-based ligands, formulating as, [Zn(valp)22,9-dmphen] (2), [Zn2(valp)4(quin)2] (3), [Zn(valp)2(2-ampy)2] (4), and [Zn(valp)2(2-ampic)2] (5) (valp = valproate, 2,9-dmphen = 2,9-dimethyl-1,10-phenanthroline, quin = quinoline, 2-ampy

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service