922749
Poly(2-methacryloyloxyethyl phosphorylcholine)
Mn 9000
Synonym(s):
PMPC
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
form
powder or crystals
Quality Level
mol wt
Mn 9000
color
white to beige
SMILES string
O=C(OCCOP(OCC[N+](C)(C)C)([O-])=O)C(C)(C)CC
Related Categories
Application
Poly (2-Methacryloyloxyethyl phosphorylcholine) (PMPC) is a methacrylate based polymer with a zwitterionic phosphorylcholine moiety on the side chain. The phosphorylcholine moiety is very similar to the head group of phospholipids in cell membranes. Due to this unique structure, PMPC showed excellent resistance to nonspecific protein adsorption, cell adhesion, and blood coagulation, as well as cell membrane penetration and underwater lubrication. PMPC has been widely exploited in a diverse range of applications, such as antifouling materials for water treatment, bioengineering, and nanomedicine.
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Critical update on 2-methacryloyloxyethyl phosphorylcholine (MPC) polymer science
Journal of Applied Polymer Science, 132, 41766-41766 (2015)
Bioconjugate chemistry, 19(11), 2144-2155 (2008-11-01)
The water-soluble, biocompatible polymer poly(2-methacryloyloxyethyl phosphorylcholine) (PMPC) was evaluated for protein conjugation. PMPC is a zwitterionic polymer that is able to form a more compact conformation in aqueous solution than poly(ethylene glycol) (PEG). While a terminally functionalized N-hydroxysuccinimide derivative of
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service