Skip to Content
Merck
All Photos(1)

Key Documents

910953

Sigma-Aldrich

di-n-butylammonium lead(II) tetrabromide

n=1

Synonym(s):

(BA)2PbBr4, (BuNH3)2PbBr4, 2D perovskite, Bis(butylammonium) tetrabromoplumbate, Butylammonium lead bromide, di-n-butylammonium tetrabromoplumbate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
(C4H9NH3)2PbBr4
CAS Number:
UNSPSC Code:
12352101

form

solid

General description

Organic-inorganic hybrid perovskittes based semiconducting materials exhibit excellent properties such as long charge carrier lifetime and diffusion length, and high photolumiscence quantum yield. These properties render the perovskite materials useful for applications in light emitting diodes (LEDs), lasers and photodetectors. The di-n-butylammonium tetrabromoplumbate is 2D-layered perovskite and finds applications as violet emitter in LEDs.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Repr. 1A - STOT RE 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dong Liang et al.
ACS nano, 10(7), 6897-6904 (2016-06-24)
Violet electroluminescence is rare in both inorganic and organic light-emitting diodes (LEDs). Low-cost and room-temperature solution-processed lead halide perovskites with high-efficiency and color-tunable photoluminescence are promising for LEDs. Here, we report room-temperature color-pure violet LEDs based on a two-dimensional lead
Giant Two-Photon Absorption and Its Saturation in 2D Organic?Inorganic Perovskite.
Liu W, et al.
Advanced Optical Materials, 5, 1601045-1601045 (2017)
Effects of Organic Moieties on Luminescence Properties of Organic-Inorganic Layered Perovskite-Type Compounds
Kawano N, et al.
The Journal of Physical Chemistry C, 118, 9101-9101 (2014)
Shuang Yang et al.
Angewandte Chemie (International ed. in English), 56(15), 4252-4255 (2017-03-16)
Two-dimensional (2D) organic-inorganic hybrid perovskite nanosheets (NSs) are attracting increasing research interest due to their unique properties and promising applications. Here, for the first time, we report the facile synthesis of single- and few-layer free-standing phenylethylammonium lead halide perovskite NSs

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service