Recommended Products
form
liquid
manufacturer/tradename
Shell
density
~1.03 g/mL at 20 °C
application(s)
detection
General description
Teepol™ 610 S is a mixture of surfactants with strong wetting and cleaning activity.
Application
Teepol™ 610 S has been used to prevent surface rippling of the solution in the kinetic study of the enzyme in carbon capture technology.
Legal Information
Teepol is a trademark of Shell Chemical Co.
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
The Journal of hygiene, 85(2), 181-191 (1980-10-01)
In a multi-laboratory trial with the membrane filtration technique, three surfactants--Teepol 610 (T610), Tergitol 7 (T7) and sodium lauryl sulphate (LS)--were compared in media for the enumeration of coliform organisms and Escherichia coli in water. A total of 179 samples
Influence of temperature and solvent concentration on the kinetics of the enzyme carbonic anhydrase in carbon capture technology
Chemical Engineering Journal, 309, 772-786 (2017)
Journal of clinical chemistry and clinical biochemistry. Zeitschrift fur klinische Chemie und klinische Biochemie, 20(10), 757-760 (1982-10-01)
Catalytic conversion of H2O2 and 2,2'-azino-di(3-ethyl-benzthiazoline-sulphonic acid-(6)) (ABTS) by peroxidase is stopped immediately and completely by the addition of a secondary alkyl sulphate (Teepol-610) or sodium dodecyl hydrogensulphate. The resulting colour intensity of the oxidized ABTS+ is thereby stabilized and
Zentralblatt fur Bakteriologie, Parasitenkunde, Infektionskrankheiten und Hygiene. Zweite naturwissenschaftliche Abteilung: Mikrobiologie der Landwirtschaft der Technologie und des Umweltschutzes, 135(2), 152-157 (1980-01-01)
Tests for susceptibility of three repesentative surface-active agents, incorporated in solid and liquid medium, were carried through with 75 strains of the Streptococcus group. Hyamine 2389 was the most active compound against the tested strains, followed by Teepol 610. Nearly
The British journal of dermatology, 116(2), 171-177 (1987-02-01)
Application of 1% potassium hydroxide (KOH) reduced subsequent development of anthralin inflammation without loss of its therapeutic effect on psoriasis. Teepol had a similar but smaller effect on subsequent development of inflammation. The action of KOH appears to have resulted
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service