Skip to Content
Merck
All Photos(1)

Documents

779679

Sigma-Aldrich

(RS)-1,1-Dimethoxy-2-propanol

≥97.0% (GC)

Synonym(s):

2-Hydroxypropionaldehyde dimethyl acetal, rac-Lactaldehyde dimethyl acetal

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H12O3
CAS Number:
Molecular Weight:
120.15
Beilstein:
1918747
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥97.0% (GC)

form

liquid

SMILES string

COC(OC)C(C)O

InChI

1S/C5H12O3/c1-4(6)5(7-2)8-3/h4-6H,1-3H3

InChI key

PRAYXKGWSGUXQK-UHFFFAOYSA-N

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

123.3 °F

Flash Point(C)

50.7 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service