Skip to Content
Merck
All Photos(1)

Key Documents

757837

Sigma-Aldrich

Poly(ethylene glycol) 2-mercaptoethyl ether acetic acid

average Mn 3,400 (n~77), cross-linking reagent amine reactive, cross-linking reagent thiol reactive

Synonym(s):

Polyethylene glycol, mercaptoPEG acid, mercaptopoly(ethylene glycol) carboxylic acid, mercaptopolyoxyethylene acetic acid, thio-PEG, thiol-PEG-acid, thiol-PEG-carboxylate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HSC2H4O(C2H4O)nCH2CO2H
CAS Number:
MDL number:
UNSPSC Code:
12162002
NACRES:
NA.23
Pricing and availability is not currently available.

Product Name

Poly(ethylene glycol) 2-mercaptoethyl ether acetic acid, average Mn 3,500

form

solid

Quality Level

mol wt

PEG average Mn 3,400 (n~77)
average Mn 3,500

reaction suitability

reagent type: cross-linking reagent
reactivity: amine reactive

reagent type: cross-linking reagent
reactivity: thiol reactive

transition temp

Tm 52-57 °C

Ω-end

thiol

α-end

carboxylic acid

polymer architecture

shape: linear
functionality: heterobifunctional

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

Application

Reported applications include: bioconjugation, drug delivery, PEG hydrogel, crosslinker, and surface functionalization.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

>604.4 °F

Flash Point(C)

> 318 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Designing biomaterial scaffolds mimicking complex living tissue structures is crucial for tissue engineering and regenerative medicine advancements.

Designing biomaterial scaffolds mimicking complex living tissue structures is crucial for tissue engineering and regenerative medicine advancements.

Designing biomaterial scaffolds mimicking complex living tissue structures is crucial for tissue engineering and regenerative medicine advancements.

Designing biomaterial scaffolds mimicking complex living tissue structures is crucial for tissue engineering and regenerative medicine advancements.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service