Skip to Content
Merck
All Photos(1)

Key Documents

724246

Sigma-Aldrich

Bisphenol A sulfate sodium salt

95% (CP)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H16O5SNa
CAS Number:
Molecular Weight:
331.34
MDL number:
UNSPSC Code:
12352108
PubChem Substance ID:

Assay

95% (CP)

form

solid

technique(s)

mass spectrometry (MS): suitable

SMILES string

[Na+].CC(C)(c1ccc(O)cc1)c2ccc(OS([O-])(=O)=O)cc2

InChI

1S/C15H16O5S.Na/c1-15(2,11-3-7-13(16)8-4-11)12-5-9-14(10-6-12)20-21(17,18)19;/h3-10,16H,1-2H3,(H,17,18,19);/q;+1/p-1

InChI key

IENSPWAUWOIJJS-UHFFFAOYSA-M

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Other Notes

Bulk stock may be available for this product. Please contact the Stable Isotope Customer Service Group at 937-859-1808 (1-800-448-9760 for US & Canada) or email us at [email protected]

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Repr. 2 - Skin Sens. 1 - STOT SE 3

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Luigi Lopardo et al.
Environment international, 125, 1-8 (2019-01-29)
Molecular epidemiology in human biomonitoring allows for verification of public exposure to chemical substances. Unfortunately, due to logistical difficulties and high cost, it evaluates only small study groups and as a result does not provide comprehensive large scale community-wide exposure

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service