Skip to Content
Merck
All Photos(1)

Key Documents

719838

Sigma-Aldrich

1-Bromo-2,6-diisopropylbenzene

95%

Synonym(s):

2,6-Diisopropylbromobenzene, 2,6-Diisopropylphenyl bromide, 2-Bromo-1,3-bis(1-methylethyl)benzene, 2-Bromo-1,3-diisopropylbenzene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H17Br
CAS Number:
Molecular Weight:
241.17
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Quality Level

Assay

95%

form

liquid

refractive index

n20/D 1.532

density

1.171 g/mL at 25 °C

functional group

bromo

SMILES string

CC(C)c1cccc(C(C)C)c1Br

InChI

1S/C12H17Br/c1-8(2)10-6-5-7-11(9(3)4)12(10)13/h5-9H,1-4H3

InChI key

QEOQKWIURDCGIJ-UHFFFAOYSA-N

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

210.0 °F - closed cup

Flash Point(C)

98.9 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service