Skip to Content
Merck
All Photos(1)

Key Documents

715123

Sigma-Aldrich

2-Chloro-6-hydroxybenzaldehyde

97%

Synonym(s):

6-Chlorosalicylaldehyde

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H5ClO2
CAS Number:
Molecular Weight:
156.57
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
form:
solid
Assay:
97%

Quality Level

Assay

97%

form

solid

mp

51-55 °C

functional group

aldehyde
chloro

storage temp.

2-8°C

SMILES string

Oc1cccc(Cl)c1C=O

InChI

1S/C7H5ClO2/c8-6-2-1-3-7(10)5(6)4-9/h1-4,10H

InChI key

MVTWVXYIKIVAOJ-UHFFFAOYSA-N

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

224.6 °F - closed cup

Flash Point(C)

107 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service