Skip to Content
Merck
All Photos(1)

Key Documents

698628

Sigma-Aldrich

1-Methyl-1H-pyrazole-5-boronic acid pinacol ester

97%

Synonym(s):

1-Methyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole, 2-(1-Methyl-1H-pyrazol-5-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-Methyl-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-2H-pyrazole

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H17BN2O2
CAS Number:
Molecular Weight:
208.07
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Assay

97%

form

powder

mp

67-71 °C

SMILES string

Cn1nccc1B2OC(C)(C)C(C)(C)O2

InChI

1S/C10H17BN2O2/c1-9(2)10(3,4)15-11(14-9)8-6-7-12-13(8)5/h6-7H,1-5H3

InChI key

HLXOVAMYQUFLPE-UHFFFAOYSA-N

Application

Reagent used for the stereoselective preparation of N-[1-(cyanomethylaminocarbonyl)-2-arylethyl]arylamides used as Cathepsin inhibitors

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Paul A Bethel et al.
Bioorganic & medicinal chemistry letters, 19(16), 4622-4625 (2009-07-21)
A number of molecular recognition features have been exploited in structure-based design of selective Cathepsin inhibitors.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service