Skip to Content
Merck
All Photos(1)

Key Documents

657425

Sigma-Aldrich

Methyl 5-bromopyridine-3-carboxylate

97%

Synonym(s):

5-Bromonicotinic acid methyl ester, 5-Bromopyridine-3-carboxylic acid methyl ester, Methyl 5-bromonicotinate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H6BrNO2
CAS Number:
Molecular Weight:
216.03
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

mp

96-100 °C

SMILES string

COC(=O)c1cncc(Br)c1

InChI

1S/C7H6BrNO2/c1-11-7(10)5-2-6(8)4-9-3-5/h2-4H,1H3

InChI key

AAJZXPWBILCHAW-UHFFFAOYSA-N

Application

Substrate used in a microwave-assisted, palladium-catalyzed arylation of acetone as its TMS enol ether.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tetrahedron Letters, 48, 1213-1213 (2007)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service