605999
L-Ascorbic acid-1-13C
99 atom % 13C
Synonym(s):
L-Threoascorbic acid-1-13C, Vitamin C-1-13C
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
isotopic purity
99 atom % 13C
Quality Level
Assay
99% (CP)
form
powder
technique(s)
mass spectrometry (MS): suitable
color
white to beige
mp
193 °C (dec.) (lit.)
mass shift
M+1
SMILES string
OC[C@H](O)[C@H]1O[13C](=O)C(O)=C1O
InChI
1S/C6H8O6/c7-1-2(8)5-3(9)4(10)6(11)12-5/h2,5,7-10H,1H2/t2-,5+/m0/s1/i6+1
InChI key
CIWBSHSKHKDKBQ-HYAXHRHPSA-N
Related Categories
General description
L-Ascorbic acid or vitamin C is a water-soluble vitamin found in various food items. It is useful in the biosynthesis of collagen, L-carnitine, and neurotransmitters. Vitamin C is also function as an antioxidant.
Application
L-Ascorbic acid-1-13C is also known as L-threoascorbic acid-1-13C. This stable isotope internal standard is suitable for the quantitation of L-ascorbic acid in dietary supplements, fortified foods or biological fluids using a variety of testing applications.
Packaging
This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
First bioarchaeological evidence of probable scurvy in Southeast Asia: multifactorial etiologies of vitamin C deficiency in a tropical environment
International Journal of Paleopathology, 5(1), 63-71 (2014)
Vitamin c
FEBS Journal, 274(1), 1-22 (2007)
A review of micronutrients in sepsis: the role of thiamine, l-carnitine, vitamin C, selenium and vitamin D
Nutrition Research Reviews, 31(2), 281-290 (2018)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service