570419
Disperse Red 1 methacrylate
95%
Synonym(s):
(4-Nitrophenyl)[4-[[2-(methacryloyloxy)ethyl]ethylamino]phenyl]diazene, 4-[(2-Methacryloxyethyl)ethylamino]-4′-nitroazobenzene
About This Item
Recommended Products
Assay
95%
form
solid
mp
78-83 °C (lit.)
λmax
493 nm
storage temp.
2-8°C
SMILES string
CCN(CCOC(=O)C(C)=C)c1ccc(cc1)\N=N\c2ccc(cc2)[N+]([O-])=O
InChI
1S/C20H22N4O4/c1-4-23(13-14-28-20(25)15(2)3)18-9-5-16(6-10-18)21-22-17-7-11-19(12-8-17)24(26)27/h5-12H,2,4,13-14H2,1,3H3/b22-21+
InChI key
MHYWMAUCJNDHPY-QURGRASLSA-N
Related Categories
General description
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
11 - Combustible Solids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service