Skip to Content
Merck
All Photos(2)

Key Documents

549541

Sigma-Aldrich

Antimony tin oxide

nanopowder, <50 nm particle size, ≥99.5% trace metals basis

Synonym(s):

ATO

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352302
NACRES:
NA.23
Pricing and availability is not currently available.

Quality Level

Assay

≥99.5% trace metals basis

form

nanopowder

composition

antimony pentoxide, 7-11%
tin(IV) oxide, 89-93%

reaction suitability

reagent type: catalyst
core: antimony

surface area

47 m2/g

particle size

<50 nm

SMILES string

O=[Sn]=O.O=[Sb]O[Sb]=O

InChI key

DCEPJBOKQZTMOG-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service