Skip to Content
Merck
All Photos(1)

Key Documents

546739

Sigma-Aldrich

Zinc bromide dihydrate

99%

Synonym(s):

Dibromozinc dihydrate, Zinc(2+) dibromide dihydrate, Zinc(II) bromide dihydrate, zinc bromide 2-hydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ZnBr2 · 2H2O
CAS Number:
Molecular Weight:
261.23
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
Assay:
99%
form:
powder or solid

Quality Level

Assay

99%

form

powder or solid

mp

>300 °C (lit.)

SMILES string

O.O.Br[Zn]Br

InChI

1S/2BrH.2H2O.Zn/h2*1H;2*1H2;/q;;;;+2/p-2

InChI key

NSGOPMOPSWFFLF-UHFFFAOYSA-L

Related Categories

Pictograms

CorrosionEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service