Skip to Content
Merck
All Photos(1)

Key Documents

540250

Sigma-Aldrich

(S)-(+)-1,2-Propanediol

96%

Synonym(s):

(S)-(+)-Propylene glycerol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH(OH)CH2OH
CAS Number:
Molecular Weight:
76.09
Beilstein:
1718871
MDL number:
UNSPSC Code:
12352001
PubChem Substance ID:
NACRES:
NA.22

Assay

96%

optical activity

[α]20/D +16.5°, neat

optical purity

ee: 98% (GLC)

refractive index

n20/D 1.432 (lit.)

bp

186-188 °C/765 mmHg (lit.)

density

1.036 g/mL at 20 °C (lit.)

functional group

hydroxyl

SMILES string

C[C@H](O)CO

InChI

1S/C3H8O2/c1-3(5)2-4/h3-5H,2H2,1H3/t3-/m0/s1

InChI key

DNIAPMSPPWPWGF-VKHMYHEASA-N

Looking for similar products? Visit Product Comparison Guide

Application

(S)-(+)-1,2-Propanediol can be used as a reactant to synthesize:
  • Optically active chiral (S)-1, 2-propanediol derivatives using mesogenic carboxylic acids via esterification in the presence of dimethylaminopyridine (DMAP) as a catalyst.
  • 3,4-Ethylenedioxythiophenes via Mitsunobu reaction.
  • Vicinal diol cyclic sulfates in the presence of sodium periodate and ruthenium chloride as catalysts.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

217.4 °F - closed cup

Flash Point(C)

103 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dae-Hee Lee et al.
Applied and environmental microbiology, 76(13), 4158-4168 (2010-05-04)
Laboratory adaptive evolution studies can provide key information to address a wide range of issues in evolutionary biology. Such studies have been limited thus far by the inability of workers to readily detect mutations in evolved microbial strains on a
Maria Pina Rosato et al.
Theriogenology, 79(3), 508-516 (2012-12-12)
This study was designed to improve current freezing protocols for rabbit sperm by examining: (1) the toxicity of different permeable cryoprotectants (CPAs) used for standard vapor freezing (conventional freezing); (2) the feasibility of ultrarapid nonequilibrium freezing (vitrification) of sperm in
R Mateus et al.
International journal of pharmaceutics, 444(1-2), 106-108 (2013-01-30)
Confocal Raman spectroscopy (CRS) is a novel approach which has been used to determine stratum corneum thickness and to profile endogenous skin components. However, the utility of this technique for probing drug disposition in vivo has not been explored to
Propylene glycol and glycerol concentration in ultrasound gel.
David Belavy et al.
Regional anesthesia and pain medicine, 38(1), 75-76 (2012-12-25)
María Gabriela Pacios et al.
Journal of investigative and clinical dentistry, 3(4), 264-270 (2012-11-07)
To evaluate the in vitro action of vehicles alone and with calcium hydroxide against different bacterial species. Agar plates were inoculated with the microbial suspensions, and wells were made and filled with the calcium hydroxide pastes and the vehicles used

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service