Skip to Content
Merck
All Photos(1)

Key Documents

516406

Sigma-Aldrich

4-Ethoxy-3-methoxybenzaldehyde

98%

Synonym(s):

4-Ethoxy-m-anisaldehyde

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C2H5OC6H3(OCH3)CHO
CAS Number:
Molecular Weight:
180.20
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

mp

50-53 °C (lit.)

SMILES string

CCOc1ccc(C=O)cc1OC

InChI

1S/C10H12O3/c1-3-13-9-5-4-8(7-11)6-10(9)12-2/h4-7H,3H2,1-2H3

InChI key

BERFDQAMXIBOHM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Demethylation of Methyl Aryl Ethers: 4-Ethoxy-3-Hydroxybenzaldehyde.
Ireland RE and Walba DM.
Organic Syntheses, 44-44 (1977)
F Cui et al.
Bioorganic & medicinal chemistry, 2(7), 735-742 (1994-07-01)
The degradation of four dimeric lignin model compounds by meso-tetra(2,6-dichloro-3-sulfonatophenyl)porphyrin iron chloride (TDCSPPFeCl) (2) are reported. 4-Ethoxy-3-methoxyphenylglycerol-beta-guaiacyl ether (3) (a beta-0-4 dimer) was cleaved to give 4-ethoxy-3-methoxybenzaldehyde (4) and guaiacol (5) as major products. The oxidation of 1-(4-ethoxy-3-methoxyphenyl)-2-(4-methoxyphenyl)-1,3-propandiol (6, a
H Wariishi et al.
The Journal of biological chemistry, 264(24), 14185-14191 (1989-08-25)
In the presence of MnII, H2O2, and glutathione (GSH), manganese peroxidase oxidized veratryl alcohol (I) to veratraldehyde (IV). Anisyl alcohol (II) and benzyl alcohol (III) were also oxidized by this system to their corresponding aldehydes (V and VI). In the
4-Ethoxy-3-methoxybenzaldehyde.
Leka Z, et al.
Acta Crystallographica Section E, Structure Reports Online, 69(12), o1728-o1728 (2013)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service