Skip to Content
Merck
All Photos(1)

Documents

486892

Sigma-Aldrich

Methyl-d3-amine

99 atom % D

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CD3NH2
CAS Number:
Molecular Weight:
34.08
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.12

vapor density

1.08 (20 °C, vs air)

Quality Level

vapor pressure

27 psi ( 20 °C)

isotopic purity

99 atom % D

autoignition temp.

806 °F

bp

−6.3 °C (lit.)

mp

−93 °C (lit.)

mass shift

M+3

SMILES string

[2H]C([2H])([2H])N

InChI

1S/CH5N/c1-2/h2H2,1H3/i1D3

InChI key

BAVYZALUXZFZLV-FIBGUPNXSA-N

Packaging

10 g quantity is packaged in a 450 mL carbon steel lecture bottle with stainless steel CGA 110/180 valve. Nominal gas pressures at 21ºC is 29 psig for 10 g. This pressure is slightly above atmospheric pressure. Care must be taken when extracting this product from the cylinder.
All gas volumes are quoted at Standard Temperature and Pressure (STP) and not at Normal Temperature and Pressure (NTP). One STP Liter is 7.7% more gas than one NTP Liter.
This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Recommended products

Stainless steel control valves Z146951, Z187224 or either of stainless steel regulators Z148504 and Z148512 and a "T"-purge valve Z151661 is recommended.
PTFE sealing washers GCA inlet size 180 Z146994
PTFE sealing washers CGA inlet size 110 Z146986

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Flam. Gas 1 - Press. Gas Liquefied gas - Skin Corr. 1B

Storage Class Code

2A - Gases

WGK

WGK 1

Flash Point(F)

<-22.0 °F - closed cup

Flash Point(C)

< -30.00 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service