Skip to Content
Merck
All Photos(2)

Key Documents

480525

Sigma-Aldrich

Lithium azide solution

20 wt. % in H2O

Synonym(s):

Lthium triazide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
LiN3
CAS Number:
Molecular Weight:
48.96
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
grade:
for analytical purposes
form:
liquid
Pricing and availability is not currently available.

grade

for analytical purposes

form

liquid

concentration

20 wt. % in H2O

density

1.088 g/mL at 25 °C

SMILES string

[Li]N=[N+]=[N-]

InChI

1S/Li.N3/c;1-3-2/q+1;-1

InChI key

GUWHRJQTTVADPB-UHFFFAOYSA-N

Application

Lithium azide can be used as:
  • A precursor in the synthesis of lithium pentazolate.
  • As an electrolyte additive for Li-S batteries to enhance their cyclability and interfacial stability.
  • As porogen reagent and nitrogen source in the preparation of nitrogen-doped porous carbon nanofibers.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - Acute Tox. 3 Dermal

Supplementary Hazards

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Since the preparation of the first organic azide, phenyl azide, by Peter Griess in 1864 this energy-rich and versatile class of compounds has enjoyed considerable interest.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service