Skip to Content
Merck
All Photos(1)

Key Documents

47578

Sigma-Aldrich

Fmoc-Asp-OAll

≥97.0% (HPLC)

Synonym(s):

Fmoc-L-aspartic acid 1-allyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H21NO6
CAS Number:
Molecular Weight:
395.41
Beilstein:
5460108
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.26
Pricing and availability is not currently available.

Quality Level

Assay

≥97.0% (HPLC)

optical activity

[α]20/D −24.5±2°, c = 1% in DMF

reaction suitability

reaction type: Fmoc solid-phase peptide synthesis

application(s)

peptide synthesis

functional group

Fmoc

storage temp.

2-8°C

SMILES string

OC(=O)C[C@H](NC(=O)OCC1c2ccccc2-c3ccccc13)C(=O)OCC=C

InChI

1S/C22H21NO6/c1-2-11-28-21(26)19(12-20(24)25)23-22(27)29-13-18-16-9-5-3-7-14(16)15-8-4-6-10-17(15)18/h2-10,18-19H,1,11-13H2,(H,23,27)(H,24,25)/t19-/m0/s1

InChI key

ZJMVIWUCCRKNHY-IBGZPJMESA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jonas Christoffersson et al.
Biofabrication, 11(1), 015013-015013 (2018-12-14)
Liver cell culture models are attractive in both tissue engineering and for development of assays for drug toxicology research. To retain liver specific cell functions, the use of adequate cell types and culture conditions, such as a 3D orientation of
Jisi Zhao et al.
Molecules (Basel, Switzerland), 24(4) (2019-03-03)
Photodynamic therapy (PDT) is an established therapeutic modality for the management of cancers. Conjugation with tumor-specific small molecule ligands (e.g., short peptides or peptidomimetics) could increase the tumor targeting of PDT agents, which is very important for improving the outcome

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service