Skip to Content
Merck
All Photos(1)

Key Documents

460958

Sigma-Aldrich

N-Methyldodecylamine

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)11NHCH3
CAS Number:
Molecular Weight:
199.38
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Quality Level

Assay

97%

refractive index

n20/D 1.44 (lit.)

bp

200 °C (lit.)

density

0.795 g/mL at 25 °C (lit.)

functional group

amine

SMILES string

CCCCCCCCCCCCNC

InChI

1S/C13H29N/c1-3-4-5-6-7-8-9-10-11-12-13-14-2/h14H,3-13H2,1-2H3

InChI key

OMEMQVZNTDHENJ-UHFFFAOYSA-N

General description

N-Methyldodecylamine is a secondary fatty acid amine. It is formed during the pyrolysis of N-(2-cyanoethyl)-N-methyldodecylamine under reduced pressure.[1]

Application

N-Methyldodecylamine may be used for the preparation of N,N,N,N,N,N-trimethyldodecylammonium bromide.[2]

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yongsoon Shin et al.
Journal of colloid and interface science, 284(1), 278-281 (2005-03-09)
We report here the self-assembly of surfactant molecules at the interface of air and the hygroscopic quaternary ammonium salt tetrabutylammonium acetate (TBAAc). Homogeneously dissolved surfactant molecules at 100 degrees C self-assemble upon contacting air due to high moisture adsorption by
Effect of the N-methylation of dodecylammonium chloride on the adsorption from its micellar solution
Aratono M, et al.
Bulletin of the Chemical Society of Japan, 61, 2773-2773 (1988)
B L Kabacoff et al.
IARC scientific publications, (57)(57), 347-352 (1984-01-01)
Nitrosation of water-soluble (diethanolamine) and oil-soluble (dodecylmethylamine and dicyclohexylamine) amines in the absence and presence of inhibitors in model anionic and non-ionic emulsions was studied. Nitrosation of diethanolamine occurred at similar rates in non-ionic and anionic emulsions. Surprisingly, dodecylmethylamine and
Kenichi Sakai et al.
Langmuir : the ACS journal of surfaces and colloids, 28(51), 17617-17622 (2012-11-20)
Rheological properties of alkyl dicarboxylic acid-alkylamine complex systems have been characterized. The complex materials employed in this study consist of an amino acid-based surfactant (dodecanoylglutamic acid, C12Glu) and a tertiary alkylamine (dodecyldimethylamine, C12DMA) or a secondary alkylamine (dodecylmethylamine, C12MA). (1)H
Erica L Bakota et al.
Rapid communications in mass spectrometry : RCM, 34(5), e8608-e8608 (2019-11-11)
While liquid chromatography/high-resolution mass spectrometry (LC/HRMS) is a versatile analytical technique, it is also sensitive to trace impurities. These impurities may come from a variety of sources, including reagents, solvents, and the sample matrix itself. Impurities in reagents may become

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service