Skip to Content
Merck
All Photos(1)

Key Documents

439797

Sigma-Aldrich

Calcium iodide

AnhydroBeads, −10 mesh, 99.999% trace metals basis

Synonym(s):

Calcium diiodide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CaI2
CAS Number:
Molecular Weight:
293.89
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
grade:
for synthesis
form:
beads
Pricing and availability is not currently available.

grade

for synthesis

product line

AnhydroBeads

Assay

99.999% trace metals basis

form

beads

impurities

≤15.0 ppm Trace Metal Analysis

particle size

−10 mesh

mp

779 °C (lit.)

SMILES string

I[Ca]I

InChI

1S/Ca.2HI/h;2*1H/q+2;;/p-2

InChI key

UNMYWSMUMWPJLR-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Legal Information

AnhydroBeads is a trademark of Sigma-Aldrich Co. LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service