Skip to Content
Merck
All Photos(1)

Documents

416444

Sigma-Aldrich

3-(Boc-amino)-1-propanol

97%

Synonym(s):

tert-Butyl N-(3-hydroxypropyl)carbamate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3COC(O)NHCH2CH2CH2OH
CAS Number:
Molecular Weight:
175.23
Beilstein:
1860807
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

liquid

reaction suitability

reagent type: cross-linking reagent

refractive index

n20/D 1.453

density

1.025 g/mL at 20 °C (lit.)

functional group

Boc
amine
hydroxyl

SMILES string

OCCCNC(OC(C)(C)C)=O

InChI

1S/C8H17NO3/c1-8(2,3)12-7(11)9-5-4-6-10/h10H,4-6H2,1-3H3,(H,9,11)

InChI key

XDJCYKMWJCYQJM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Amine protected, difunctional reagent employed in the synthesis of phosphatidyl ethanolamines and ornithine.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

228.2 °F - closed cup

Flash Point(C)

109 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Synthetic Communications, 21, 359-359 (1991)
Synthesis, 133-133 (1993)
Lan Yao et al.
Polymers, 12(5) (2020-05-28)
Polymeric nanomaterials made from amphiphilic block copolymers are increasingly used in the treatment of tumor tissues. In this work, we firstly synthesized the amphiphilic block copolymer PBnMA-b-P(BAPMA-co-PEGMA) via reversible addition-fragmentation chain transfer (RAFT) polymerization using benzyl methacrylate (BnMA), poly (ethylene

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service