Skip to Content
Merck
All Photos(2)

Documents

408255

Sigma-Aldrich

Neopentyl glycol diacrylate

Synonym(s):

(2,2-Dimethyl-3-prop-2-enoyloxypropyl) prop-2-enoate, 2,2-Dimethylpropanediol diacrylate, 2-Propenoic acid, 2,2-dimethyl-1,3-propanediyl ester, Neopentanediol diacrylate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(H2C=CHCO2CH2)2C(CH3)2
CAS Number:
Molecular Weight:
212.24
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

form

liquid

contains

225 ppm monomethyl ether hydroquinone as inhibitor

refractive index

n20/D 1.453 (lit.)

density

1.031 g/mL at 25 °C (lit.)

SMILES string

CC(C)(COC(=O)C=C)COC(=O)C=C

InChI

1S/C11H16O4/c1-5-9(12)14-7-11(3,4)8-15-10(13)6-2/h5-6H,1-2,7-8H2,3-4H3

InChI key

MXFQRSUWYYSPOC-UHFFFAOYSA-N

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lokendra P Singh et al.
Physical chemistry chemical physics : PCCP, 11(25), 5110-5118 (2009-06-30)
In the present article, investigations of an unusual two-component (H-) bonded pair, i.e. the cyclohexanol-neopentylglycol system, are reported. The phase I of cyclohexanol (CHXOL) forms a continuous solid solution with the phase I of neopentylglycol (NPGOL). This binary solid solution
B Björkner et al.
Contact dermatitis, 11(5), 268-278 (1984-11-01)
Many factors can influence the elicitation of hypersensitivity reactions in guinea pigs and humans. The effect which the vehicle might have on the test response in guinea pigs sensitized with various acrylic compounds, using the "guinea pig maximization test", has
Y Abe et al.
Journal of oral rehabilitation, 28(5), 407-412 (2001-06-13)
This in vitro study evaluated the wear effects of five posterior denture tooth materials on human enamel. The tooth specimen was cusp shaped and enamel specimen was formed as a 10 C 10 C 5 mm plate. All material-enamel combinations

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service