Skip to Content
Merck
All Photos(1)

Key Documents

391654

Sigma-Aldrich

(1R,2R,3R,5S)-(−)-Isopinocampheylamine

95%

Synonym(s):

(−)-Isopinocampheylamine, (1R,2R,3R,5S)-3-Pinanamine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H19N
CAS Number:
Molecular Weight:
153.26
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Quality Level

Assay

95%

form

liquid

optical activity

[α]21/D −42°, neat

refractive index

n20/D 1.48 (lit.)

bp

90 °C/18 mmHg (lit.)

density

0.909 g/mL at 25 °C (lit.)

SMILES string

C[C@H]1[C@H](N)CC2CC1C2(C)C

InChI

1S/C10H19N/c1-6-8-4-7(5-9(6)11)10(8,2)3/h6-9H,4-5,11H2,1-3H3/t6-,7+,8-,9-/m1/s1

InChI key

VPTSZLVPZCTAHZ-BZNPZCIMSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

161.6 °F - closed cup

Flash Point(C)

72 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Wenhui Hu et al.
Journal of medicinal chemistry, 53(9), 3831-3834 (2010-04-17)
Although amantadine derivatives are the only M2 drugs for influenza virus A, their use is limited in the U.S. because of drug resistance. Here we report the identification of multiple M2 inhibitors that were rapidly generated through focused screening of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service