Skip to Content
Merck
All Photos(1)

Key Documents

37278

Sigma-Aldrich

(−)-Dihydrocarveol

mixture of isomers, ≥95.0% (sum of enantiomers, GC)

Synonym(s):

(1R,2R,5R)-5-Isopropenyl-2-methylcyclohexanol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H18O
CAS Number:
Molecular Weight:
154.25
Beilstein:
2433199
MDL number:
UNSPSC Code:
12352002
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Assay

≥95.0% (sum of enantiomers, GC)

optical activity

[α]20/D −20±1°, neat

composition

n-, ~75%
iso-, ~6%
neo-, ~3%
neoiso-, ~13%

refractive index

n20/D 1.476-1.479

density

0.926 g/mL at 20 °C (lit.)

SMILES string

C[C@@H]1CC[C@H](C[C@H]1O)C(C)=C

InChI

1S/C10H18O/c1-7(2)9-5-4-8(3)10(11)6-9/h8-11H,1,4-6H2,2-3H3/t8-,9-,10-/m1/s1

InChI key

KRCZYMFUWVJCLI-OPRDCNLKSA-N

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

194.0 °F - closed cup

Flash Point(C)

90 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service