365483
(Methoxycarbonylsulfamoyl)triethylammonium hydroxide, inner salt
97%
Synonym(s):
Burgess reagent, Methyl N-(triethylammoniosulfonyl)carbamate
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Linear Formula:
CH3O2CNSO2N(C2H5)3
CAS Number:
Molecular Weight:
238.30
Beilstein:
1432131
MDL number:
UNSPSC Code:
12352108
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Quality Level
Assay
97%
form
powder
mp
76-79 °C (lit.)
functional group
amine
storage temp.
−20°C
SMILES string
CC[N+](CC)(CC)S(=O)(=O)[N-]C(=O)OC
InChI
1S/C8H18N2O4S/c1-5-10(6-2,7-3)15(12,13)9-8(11)14-4/h5-7H2,1-4H3
InChI key
YSHOWEKUVWPFNR-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Application
Powerful dehydration agent for compounds such as secondary and tertiary alcohols, diols, amino alcohols, sugars, etc., including reactions with epoxides.
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Synlett, 9, 1247-1247 (2003)
K C Nicolaou et al.
Journal of the American Chemical Society, 126(20), 6234-6235 (2004-05-20)
Although glycosylamines constitute an important group of carbohydrates from the standpoint of biology and medicine, methods for their synthesis typically lack substrate generality and/or result in variable stereoselectivity, especially in complex contexts. In this communication, we report an operationally simple
A A Nagel et al.
Journal of medicinal chemistry, 25(7), 881-884 (1982-07-01)
Ring contraction of the neutral oleandrose sugar in the 14-membered-ring macrolide antibiotic oleandomycin (2) has been accomplished using [(methoxycarbonyl)sulfamoyl]triethylammonium hydroxide inner salt (1). The product of this interesting rearrangement, after methanolic hydrolysis of the 2'-acetate, is the 11-acetyl-3-O-(3"-methoxy-4"-vinylfuranosyl)oleandomycin (12). The
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service