Skip to Content
Merck
All Photos(1)

Documents

362999

Sigma-Aldrich

Benzyl-2,3,4,5,6-d5 alcohol

98 atom % D

Synonym(s):

(2,3,4,5,6-Pentadeuteriophenyl)methanol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6D5CH2OH
CAS Number:
Molecular Weight:
113.17
MDL number:
UNSPSC Code:
12352001
PubChem Substance ID:
NACRES:
NA.12

isotopic purity

98 atom % D

Quality Level

Assay

99% (CP)

form

liquid

refractive index

n20/D 1.539 (lit.)

bp

203-205 °C (lit.)

mp

-16--13 °C (lit.)

density

1.094 g/mL at 25 °C

mass shift

M+5

SMILES string

[2H]c1c([2H])c([2H])c(CO)c([2H])c1[2H]

InChI

1S/C7H8O/c8-6-7-4-2-1-3-5-7/h1-5,8H,6H2/i1D,2D,3D,4D,5D

InChI key

WVDDGKGOMKODPV-RALIUCGRSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

204.8 °F - closed cup

Flash Point(C)

96 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Amélie Slegers et al.
Molecules (Basel, Switzerland), 20(6), 10980-11016 (2015-06-18)
Developed from crosses between Vitis vinifera and North American Vitis species, interspecific hybrid grape varieties are becoming economically significant in northern areas, where they are now extensively grown for wine production. However, the varietal differences between interspecific hybrids are not

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service