359181
Tetraethyl methylenediphosphonate
97%
Synonym(s):
Bis(diethoxyphosphinyl)methane, Methylenediphosphonic acid tetraethylester, NSC 133889
About This Item
Recommended Products
Assay
97%
form
liquid
reaction suitability
reaction type: C-C Bond Formation
refractive index
n20/D 1.442 (lit.)
bp
171-174 °C/11 mmHg (lit.)
density
1.16 g/mL at 25 °C (lit.)
functional group
phosphonate
SMILES string
CCOP(=O)(CP(=O)(OCC)OCC)OCC
InChI
1S/C9H22O6P2/c1-5-12-16(10,13-6-2)9-17(11,14-7-3)15-8-4/h5-9H2,1-4H3
InChI key
STJWVOQLJPNAQL-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
Application
- Dual substrate-site inhibitors of 3-deoxy-D-arabino-heptulosonate 7-phosphate synthase
- Unnatural alpha-amino acid derivatives containing gem-biphosphonates
- Biphenyl sulfonylamino Me bisphosphonic acids as inhibitors of matrix metalloproteinases and bone resorption
- Lycopene via Wittig-Horner reaction
- Alkylaminoethylbisphosphinic acids to target farnesyl diphosphate synthase
- Aromatic bisphosphonates for use as inhibitors of geranylgeranyl diphosphate synthase
Precursor for synthesis of dendritic polyglycerol anions used toward L-selectin inhibition
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Flash Point(F)
230.0 °F - closed cup
Flash Point(C)
110 °C - closed cup
Personal Protective Equipment
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service