Skip to Content
Merck
All Photos(2)

Key Documents

326844

Sigma-Aldrich

1,1,1-Tris(4-hydroxyphenyl)ethane

99%

Synonym(s):

4,4′,4′′-Ethylidynetrisphenol, THPE

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C(C6H4OH)3
CAS Number:
Molecular Weight:
306.36
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

Assay

99%

mp

246-248 °C (lit.)

SMILES string

CC(c1ccc(O)cc1)(c2ccc(O)cc2)c3ccc(O)cc3

InChI

1S/C20H18O3/c1-20(14-2-8-17(21)9-3-14,15-4-10-18(22)11-5-15)16-6-12-19(23)13-7-16/h2-13,21-23H,1H3

InChI key

BRPSWMCDEYMRPE-UHFFFAOYSA-N

Pictograms

Environment

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Chien-Shu Chen et al.
Journal of medicinal chemistry, 52(9), 2716-2723 (2009-05-08)
Hepatitis C virus nonstructural protein 3 (HCV NS3) helicase is believed to be essential for viral replication and has become an attractive target for the development of antiviral drugs. A fluorescence resonant energy transfer helicase assay was established for fast

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service