292192
2-Phenyltoluene
97%
Synonym(s):
2-Methylbiphenyl
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
C6H5C6H4CH3
CAS Number:
Molecular Weight:
168.23
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Assay
97%
refractive index
n20/D 1.591 (lit.)
bp
255.3 °C (lit.)
density
1.011 g/mL at 25 °C (lit.)
functional group
phenyl
SMILES string
Cc1ccccc1-c2ccccc2
InChI
1S/C13H12/c1-11-7-5-6-10-13(11)12-8-3-2-4-9-12/h2-10H,1H3
InChI key
ALLIZEAXNXSFGD-UHFFFAOYSA-N
Related Categories
General description
Density functional theory calculations for the radical anions of 2-phenyltoluene (2-methylbiphenyl) has been reported. Cyclodehydrogenation of 2-phenyltoluene (2-methylbiphenyl) and its conversion to fluorene nucleus by vapor-phase treatment with palladium catalyst has been reported.
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Flash Point(F)
224.6 °F - closed cup
Flash Point(C)
107 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Dandan Xiang et al.
Chemosphere, 201, 840-849 (2018-03-20)
Mounting evidence suggests there is a link between exposure to synthetic pyrethroids (SPs) and the development of obesity. The information presented in this study suggests that cis-bifenthrin (cis-BF) could activate pregnane X receptor (PXR) mediated pathway and lead to the
Aromatic Cyclodehydrogenation. III. Experiments with 2-Ethylbiphenyl1
Orchin M.
Journal of the American Chemical Society, 68(4), 571-572 (1946)
Investigation of the effects of substitution position on the radical anions of chlorobiphenyls.
Pan D and Phillips DL.
Chemical Physics Letters, 318(1), 214-221 (2000)
Philipp Denk et al.
Analytical and bioanalytical chemistry, 410(14), 3395-3404 (2018-04-07)
Elastic therapeutic tapes are an important tool in the field of physical therapy and medicine. These tapes contain types of adhesive. However, sensory evaluations revealed the release of pronounced and irritating odors of the tapes. Negative odors were, amongst others
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service